Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ UTX (KDM6A), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1865MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1865MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1865MI Supplier MilliporeSigma™ Supplier No. ABE1865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysine-specific demethylase 6A (UniProt O15550; also known as Histone demethylase UTX, KDM6A, Ubiquitously-transcribed TPR protein on the X chromosome, Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein) is encoded by the KDM6A (also known as KABUK2, UTX) gene (Gene ID 7403) in human. KDM6A specifically demethylates Lys-27 of histone H3 and plays a central role in histone code. It is shown to demethylate trimethylated and dimethylated, but not monomethylated H3 Lys-27. KDM6A plays a vital role in regulation of posterior development, by regulating HOX gene expression. KDM6A also plays a potential role in regulating brown adipocyte thermogenic program. It is up-regulated during brown adipocyte differentiation and by cold exposure in both brown and white adipose tissue in mice, indicating its potential role in thermogenesis. Inactivation of KDM6A down-regulates brown fat specific gene expression. Mutations in KDM6A gene are a causative factor in Kabuki syndrome that is characterized by congenital mental retardation, postnatal dwarfism, large prominent earlobes, a cleft or high-arched palate, scoliosis, and other abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTX (KDM6A)
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene KDM6A, KABUK2, UTX
Gene Accession No. O15550
Host Species Rabbit
Immunogen His-tagged recombinant fragment within the central region of human UTX (Seenundun, S., et al. (2010). EMBO J. 29(8):1401-1411).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_066963
Test Specificity This polyclonal antibody detected the expected UTX target band in wild-type, but not Utx-knockout murine ESC nuclear extract (Faralli, H., et al. (2016). J. Clin. Invest. 126(4):1555-1565).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.