Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UTY Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526082
Description
UTY Polyclonal Antibody for Western Blot, ICC/IF, Flow
UTY encodes a protein containing tetratricopeptide repeats which are thought to be involved in protein-protein interactions. This protein is a minor histocompatibility antigen which may induce graft rejection of male stem cell grafts.
Specifications
| UTY | |
| Polyclonal | |
| Unconjugated | |
| UTY | |
| histone demethylase UTY; Hydb; Kdm6a; KDM6AL; KDM6C; lysine (K)-specific demethylase 6A; lysine-specific demethylase 6A; male-specific histocompatibility antigen H-YDB; mKIAA4057; RGD1565481; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat containing, Y-linked; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR gene on Y chromosome; ubiquitously transcribed TPR protein on the Y chromosome; ubiquitously transcribed Y chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; Utx; Uty; UTY1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7404 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O14607 | |
| UTY | |
| KLH conjugated synthetic peptide between 1247-1276 amino acids from the C-terminal region of human UTY. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction