Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568440 Supplier Invitrogen™ Supplier No. PA568440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UTY Polyclonal Antibody for Western Blot, IHC (P)

UTY encodes a protein containing tetratricopeptide repeats which are thought to be involved in protein-protein interactions. This protein is a minor histocompatibility antigen which may induce graft rejection of male stem cell grafts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTY
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UTY
Gene Accession No. O14607, P79457
Gene Alias histone demethylase UTY; Hydb; Kdm6a; KDM6AL; KDM6C; lysine (K)-specific demethylase 6A; lysine-specific demethylase 6A; male-specific histocompatibility antigen H-YDB; mKIAA4057; RGD1565481; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat containing, Y-linked; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR gene on Y chromosome; ubiquitously transcribed TPR protein on the Y chromosome; ubiquitously transcribed Y chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; Utx; Uty; UTY1
Gene Symbols UTY
Host Species Rabbit
Immunogen Full length Fusion protein of human UTY.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22290, 7404
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.