Learn More
Honeywell™ Uvex™ Astrospec 3000™ CB (Custom Bridge) Safety Glasses
Specifications
Certifications/Compliance | Meets ANSI Z87.1+ standards. |
---|---|
Lens Tint | Clear |
Frame Color | Black |
Catalog Number | Mfr. No. | Lens Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Coating | Price | Quantity & Availability | |||||
19-076070
|
Honeywell™
S2200C |
Anti-fog | N/A | N/A | N/A | ||||
Description
Custom fit for comfortable protection
Designed specifically for wearers with low-profile nasal area or large-profile cheekbone area, Astrospec 3000 CB spectacles offer features of Astrospec 3000 spectacles plus a customized lens and nose bridge.
Spectacles are lightweight and impact-resistant, with polycarbonate sideshields and browguard for protection against UV, dust, and flying particles. Black frames available with stratch-resistant Ultradura™ coating or Uvextreme™ (XTR) coating for optimal protection from harmful UV light, scratches, fogging, and static.
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Compliance
Both lens coatings meet ANSI Z87.1+ standards.
Specifications
Meets ANSI Z87.1+ standards. | |
Clear | |
Black |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.