Learn More
Honeywell™ Uvex™ Astrospec 3000™ Safety Glasses with Duoflex Temples
Catalog No. 1798531J Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Blue
Red, White, Blue
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Clear
Gray
SCT-Blue
Shade 2.0 Infra-dura
Silver Mirror
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
17-985-31J Red, White, Blue Scratch-resistant Gray
18-999-2554B Black - Clear
19-025334 Red, White, Blue Anti-fog Clear
17-985-30J Red, White, Blue Scratch-resistant Clear
18-999-2554A Black Scratch-resistant Clear
19-003-319 Black - Shade 2.0 Infra-dura
19-041-036 Black Anti-fog SCT-Blue
18-999-2553A Blue - Clear
18-999-2553B Blue Anti-fog Clear
18-999-2554D Black - Silver Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 17-985-31J Supplier Honeywell™ Supplier No. S1179
This item is not returnable. View return policy
Compliance
ANSI™ Z87+ and CSA Z94.3 standards
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87+ and CSA Z94.3 standards
Lens Tint Gray
Frame Color Red, White, Blue
Temple Style Duoflex