Learn More
Honeywell™ Uvex™ Bandit™ Safety Glasses Replacement Lenses
Catalog No. 19003338A
Click to view available options
Lens Tint:
Amber UD
Clear UD
Clear XTR
Espresso UD
Mirror UD
Contemporary styling, sporty wrap-around temples provide non-slip fit and comfort. Pack of 10 single lenses for use with easy lens replacement system.
  • Sleek, sporty design features a single polycarbonate lens with sideshields and browguards molded into a lightweight, wraparound frame.
  • High-visibility lens is available in a variety of tints, and the patented lens replacement system enables quick, easy lens changes to meet any work application.
  • Lens absorbs 99.9% of harmful UV light and is coated with scratch-resistant Ultradura™ (UD) hardcoat.
  • Clear lenses are also available with exclusive Uvextreme™ AF (XTR) antifog coating that provides additional UV protection and minimizes static and scratching.
TRUSTED_SUSTAINABILITY
Specifications
Lens Type UD
Description Replacement lens
Lens Tint Clear UD
Product Type Replacement lens
For Use With (Equipment) Bandit Spectacles
No. per Pack 200/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.