Learn More
Honeywell™ Uvex™ Carbonvision™ Safety Goggles
Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
Specifications
| Lens Coating | Scratch-resistant |
|---|---|
| Lens Type | Dura-streme HC/AF |
| Certifications/Compliance | ANSI Z87.1-2010 and CSA Z94.3 standards |
| Frame Color | Black, Gray |
| Catalog Number | Mfr. No. | Frame Material | Lens Tint | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Frame Material | Lens Tint | Price | Quantity & Availability | ||||
|
19-111-203
|
Honeywell™
S1651D |
Neoprene | Gray | N/A | N/A | N/A | |||
|
19-111-204
|
Honeywell™
S1650DF |
Fabric | Clear | N/A | N/A | N/A | |||
|
19-111-202
|
Honeywell™
S1650D |
Neoprene | Clear | N/A | N/A | N/A | |||
Description
Weighing in at just over an ounce and a half, Uvex Carbonvision goggles are one of the lightest wide vision goggles on the market. Now wearers can work for longer periods without the distractions and problems caused by discomfort, thereby increasing productivity and promoting a safer workplace.
Key Features and Benefits
- Flexible soft frame adapts perfectly to the face to ensure a perfect fit and seal around the eyes for high-performance protection and all-day comfort
- Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
- Meets the ANSI Z87.1-2010 standard
- Certified to the requirements of the CSA Z94.3 standard
For more information, visit www.fishersci.com and search for keywords UVEX Goggles.
Specifications
| Scratch-resistant | |
| Dura-streme HC/AF | |
| ANSI Z87.1-2010 and CSA Z94.3 standards | |
| Black, Gray |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.