Learn More
Honeywell™ Uvex™ Genesis™ Protective Safety Glasses
Catalog No. 19040158 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Earth
Red, White, Blue
Vapor Blue
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Clear
Espresso
Gray
IR Filter Shade 5.0
Mirror
SCT-Blue
SCT-Reflect 50
SCT-Vermillion
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-040-158 Earth Scratch-resistant Amber
19-040-143 Black Scratch-resistant Clear
19-040-145 Black Scratch-resistant Espresso
19-040-152 Black Scratch-resistant SCT-Reflect 50
19-040-168 Vapor Blue Scratch-resistant Clear
19-185-703 Vapor Blue Anti-fog Clear
19-120-1187 Red, White, Blue - Clear
17-377-417 Black Anti-fog, Scratch-resistant Clear
19-003-284 Black - IR Filter Shade 5.0
19-040-144 Black Anti-fog Clear
19-040-146 Black Anti-fog Espresso
19-040-147 Black Scratch-resistant Amber
19-040-169 Vapor Blue Anti-fog Clear
19-040-172 Vapor Blue Scratch-resistant Amber
19-040-191 Black - SCT-Blue
19-040-173 Vapor Blue Scratch-resistant Mirror
19-040-174 Vapor Blue Scratch-resistant SCT-Reflect 50
19-040-190 Black - SCT-Vermillion
19-073-200 Black Anti-fog Gray
19-820-987 Black Anti-fog Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Catalog No. 19-040-158 Supplier Honeywell™ Supplier No. S3222
This item is not returnable. View return policy
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
  • Impact-resistant polycarbonate lenses block >99.9% of harmful UVA and UVB radiation
  • Ventilation channel between frame and lens prevents moisture buildup
  • Nose bridge is covered with small, ultrasoft “fingers” that hug the nose for improved fit and comfort
  • Soft elastomer browguard cushions all points of contact with face
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
Hydroshield™ Anti-Fog Coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
XC Series
  • Extended wrap-around lens for extra cheek and extra side protection
    • Soft flexible nose fingers conform to fit a variety of nasal profiles
    • Adjustable temples and lens inclination
    ultra-dura™ Hard Coat (UD)
    • Permanently bonded to lens
    • Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meets ANSI Z87+ and CSA Z94.3 standards
Description Lifetime frame guarantee
Lens Tint Amber
Frame Color Earth