Learn More
Honeywell™ Uvex™ Genesis™ Protective Safety Glasses
Click to view available options
Frame Color:
Black
Earth
Red, White, Blue
Vapor Blue
Lens Tint:
Amber
Clear
Espresso
Gray
IR Filter Shade 5.0
Mirror
SCT-Blue
SCT-Reflect 50
SCT-Vermillion
Certifications/Compliance:
ANSI Z87.1-2015, CSA Z94.3-2007
ANSI Z87.1-2015, CSA Z94.3-2007, Military V0 Ballistic Test
Meets ANSI Z87+ and CSA Z94.3 standards
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Scratch-resistant
Description
- Impact-resistant polycarbonate lenses block >99.9% of harmful UVA and UVB radiation
- Ventilation channel between frame and lens prevents moisture buildup
- Nose bridge is covered with small, ultrasoft “fingers” that hug the nose for improved fit and comfort
- Soft elastomer browguard cushions all points of contact with face
uvextreme™ Anti-Fog Coating (XTR)
- Permanently bonded to lens
- Absorbs 99.9% of UV radiation
- Anti-scratch and -static
- Will not wear after repeated cleanings
- Provides 99.9% ultra-violet protection
- Provides 2X greater scratch-resistance
- Extended wrap-around lens for extra cheek and extra side protection
- Soft flexible nose fingers conform to fit a variety of nasal profiles
- Adjustable temples and lens inclination
- Permanently bonded to lens
- Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
Specifications
Specifications
Description | Lifetime frame guarantee |
Lens Tint | IR Filter Shade 5.0 |
Frame Color | Black |