Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Catalog No. 18813394
Click to view available options
Lens Type:
Ultra-dura Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
This item is not returnable. View return policy
Catalog No. 18-813-394
Supplier: Honeywell™ S3764
This item is not returnable. View return policy
High-impact protection
  • Soft, flexible nasal fingers
  • Padded temple tips and cushioned browbar
  • Adjustable temple length and ratcheting lens inclination provides for secure fit
  • Easy and economical lens replacement (see Cat. No. 18-813-424)
  • With Ultra-dura™ antiscratch coating
  • Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Type Ultra-dura Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2007
Lens Tint Clear
Product Type Reading Magnifier Spectacles
For Use With (Application) Ideal for most indoor work applications
Frame Color Black
Magnification Power +3.00
UV Protection UV400
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.