Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Catalog No. 18813393 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Type:
Ultra-dura Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Type Lens Tint Magnification Power
18-813-393 Ultra-dura Scratch-resistant Clear +2.50
18-813-390 Ultra-dura Scratch-resistant Clear +1.00
18-813-402 Ultra-dura Scratch-resistant Gray +2.00
18-813-403 Ultra-dura Scratch-resistant Gray +2.50
18-813-391 Ultra-dura Scratch-resistant Clear +1.50
18-813-392 Ultra-dura Scratch-resistant Clear +2.00
18-813-394 Ultra-dura Scratch-resistant Clear +3.00
18-813-395 Ultra-dura Scratch-resistant Amber +1.00
18-813-404 Ultra-dura Scratch-resistant Gray +3.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-813-393 Supplier Honeywell™ Supplier No. S3763
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-impact protection
  • Soft, flexible nasal fingers
  • Padded temple tips and cushioned browbar
  • Adjustable temple length and ratcheting lens inclination provides for secure fit
  • Easy and economical lens replacement (see Cat. No. 18-813-424)
  • With Ultra-dura™ antiscratch coating
  • Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
Specifications
Lens Coating Scratch-resistant
Lens Type Ultra-dura Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2007
Lens Tint Clear
Product Type Reading Magnifier Spectacles
For Use With (Application) Ideal for most indoor work applications
Frame Color Black
Magnification Power +2.50
UV Protection UV400