Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Shop All Protective Industrial Products Inc. ProductsDescription
- Soft, flexible nasal fingers
- Padded temple tips and cushioned browbar
- Adjustable temple length and ratcheting lens inclination provides for secure fit
- Easy and economical lens replacement (see Cat. No. 18-813-424)
- With Ultra-dura™ antiscratch coating
- Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
Specifications
Specifications
| Lens Type | Ultra-dura Scratch-resistant |
| Certifications/Compliance | ANSI Z87.1-2010 and CSA Z94.3 standards |
| Lens Tint | Gray |
| Product Type | Reading Magnifier Spectacles |
| For Use With (Application) | Minimizes outdoor sunlight and glare |
| Frame Color | Black |
| Magnification Power | +3.00 |
| UV Protection | 0.999 |