Learn More
Honeywell Safety Products™ Uvex™ Genesis™ Safety Eyewears
Shop All Protective Industrial Products Inc. ProductsDescription
- With Hydroshield™ anti-fog lens coating
- Flexible fingers adjust to most nasal profiles
- Elastomer browguard deflects impact
- Adjustable temples and lens inclination
- Provides 99.9% ultra-violet protection
- Provides 2X greater scratch-resistance
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Type | HydroShield AF |
| Lens Tint | SCT-Low IR |
| Frame Color | Black |