Learn More
Honeywell™ Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Catalog No. 19073203
Click to view available options
Description:
Replacement Lenses/Spectacles
Replacement lens
Safety Spectacles
Lens Coating:
Anti-fog
Hardcoat
Lens Tint:
Amber
Clear
Gold Mirror
Gray (50%)
SCT-Low IR
Shade 3
This item is not returnable. View return policy
Catalog No. 19-073-203
Supplier: Honeywell™ S6913X
This item is not returnable. View return policy
Wrap-around lens provides exceptional peripheral vision and protection. Flexible fingers adjust to most nasal profiles. Elastomer browguard deflects impact. Adjustable temples and lens inclination.
Uvex Genesis is engineered to deliver on the best all-day comfort, protection and style. It's built with MMT for comfort, and protects in a very wide assortment of lens tints. Uvex Genesis has an easy and economical lens replacement system. Frames have a lifetime guarantee. Made in the USA.
  • PC lens, Hydroshield AF, Dura-streme AF/HC, Uvextreme
  • Ratchet, adjustable temples, flexible nose, MMT
  • Available in a very wide range of lens tints
  • Economical lens replacement system
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type Uvextreme Anti-fog
Description Replacement lens
Lens Tint Gray (50%)
Product Type Replacement Lens
For Use With (Equipment) genesis Spectacles
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.