Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Uvex™ Genesis X2™ Safety Glasses: Replacement Lenses

Replacement lenses for Uvex Genesis X2 safety glasses (cat. no. 17377422)

Specifications

Description Replacement lenses for Uvex Genesis X2 spectacles
Product Type Replacement lens
For Use With (Equipment) Uvex Genesis X2 spectacles
No. per Pack 10/Pk.
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Application) Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Lens Tint Price Quantity & Availability  
17-377-416
View Documents
Honeywell™
S6924
Light mirror coating for varying light conditions SCT-Reflect 50 N/A N/A N/A
Description

Description

Uvextreme™ Antifog Coating (XTR)

  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
Ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant

Compliance

Exceed ANSI™ Z87.1-2010 standards. Meet stringent military V0 ballistic impact requirements.

Warranty and Services

Lifetime frame guarantee

Specifications

Specifications

Replacement lenses for Uvex Genesis X2 spectacles
Replacement lens
Uvex Genesis X2 spectacles
10/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.