Learn More
Honeywell™ Uvex™ Genesis XC™ Protective Eyewear
Shop All Protective Industrial Products Inc. ProductsDescription
- Soft, flexible nose fingers conform to fit a variety of nasal profiles
- Black frames with adjustable temples for comfortable fit; cushioned for all-day comfort
- Available lens tints include clear, gray, mirror, Infradura™ welding shades, and SCT™ to reduce spectral hazards
- All clear lenses absorb >99.9% of UVA and UVB radiation up to 385nm; tinted lenses absorb >99.9% UVA and UVB radiation up to 400nm
- Rx Insert available (see cat. no. 191300825)
uvextreme™ Anti-Fog Coating (XTR)
- Permanently bonded to lens
- Absorbs 99.9% of UV radiation
- Anti-scratch and -static
- Will not wear after repeated cleanings
- Permanently bonded to lens
- Scratch- and chemical-resistant
- ultra-dura coated exterior; uvextreme coated interior
- Lengths lens life
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
- Reduces glare for workers moving between indoor to outdoor work environments.
- Provide broad protection against accidental arcs. Equivalent to a Shade 2.0. welding filter, but not designed for any welding applications.
- Provide protection from short-term electric arcs and radiation generated when an arc accidentally occurs. Equivalent to a Shade 1.2 welding lens, but not designed for any welding applications.
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | Meet ANSI Z87.1-2003 and CSA Z94.3 standards. |
| Lens Tint | SCT-Reflect 60 |
| Frame Color | Black |
| Temple Style | Cushioned |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.