Learn More
Honeywell™ Uvex™ Genesis XC™ Protective Eyewear
Catalog No. 19066038 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Clear
Gray
IR Filter Shade 2.0
Mirror
SCT-Gray
SCT-Reflect 60
SCT-Vermilion
Shade 3
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-066-038 Scratch-resistant IR Filter Shade 2.0
19-066-055 Scratch-resistant Clear
19-185-707 Anti-fog Shade 3
19-185-709 Anti-fog Gray
19-185-710 Anti-fog SCT-Vermilion
17-377-418 Anti-fog, Scratch-resistant Clear
19-066-029 Anti-fog Clear
19-066-031 Scratch-resistant Gray
19-066-035 Scratch-resistant SCT-Reflect 60
19-066-036 Anti-fog SCT-Gray
19-066-041 Scratch-resistant Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-066-038 Supplier Honeywell™ Supplier No. S3305
This item is not returnable. View return policy
Extended wraparound lens provides excellent coverage
  • Soft, flexible nose fingers conform to fit a variety of nasal profiles
  • Black frames with adjustable temples for comfortable fit; cushioned for all-day comfort
  • Available lens tints include clear, gray, mirror, Infradura™ welding shades, and SCT™ to reduce spectral hazards
  • All clear lenses absorb >99.9% of UVA and UVB radiation up to 385nm; tinted lenses absorb >99.9% UVA and UVB radiation up to 400nm
  • Rx Insert available (see cat. no. 191300825)
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
dura-streme™ Combination Coating (DS)
  • ultra-dura coated exterior; uvextreme coated interior
  • Lengths lens life
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
  1. Reduces glare for workers moving between indoor to outdoor work environments.
  2. Provide broad protection against accidental arcs. Equivalent to a Shade 2.0. welding filter, but not designed for any welding applications.
  3. Provide protection from short-term electric arcs and radiation generated when an arc accidentally occurs. Equivalent to a Shade 1.2 welding lens, but not designed for any welding applications.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
Description Shade 2.0 Infra-dura
Lens Tint IR Filter Shade 2.0
Frame Color Black
Temple Style Cushioned