Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex™ Genesis XC™ Protective Eyewear
Extended wraparound lens provides excellent coverage
Supplier: Honeywell™ S3312HS
This item is not returnable.
View return policy
Description
- Soft, flexible nose fingers conform to fit a variety of nasal profiles
- Black frames with adjustable temples for comfortable fit; cushioned for all-day comfort
- Available lens tints include clear, gray, mirror, Infradura™ welding shades, and SCT™ to reduce spectral hazards
- All clear lenses absorb >99.9% of UVA and UVB radiation up to 385nm; tinted lenses absorb >99.9% UVA and UVB radiation up to 400nm
- Rx Insert available (see cat. no. 191300825)
uvextreme™ Anti-Fog Coating (XTR)
- Permanently bonded to lens
- Absorbs 99.9% of UV radiation
- Anti-scratch and -static
- Will not wear after repeated cleanings
- Permanently bonded to lens
- Scratch- and chemical-resistant
- ultra-dura coated exterior; uvextreme coated interior
- Lengths lens life
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.

Specifications
Anti-fog | |
SCT-Blue | |
Black | |
Adjustable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction