Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex™ Genesis XC™ Protective Eyewear
Extended wraparound lens provides excellent coverage
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Price | Quantity & Availability | ||||
19-066-038
|
Honeywell™
S3305 |
Scratch-resistant | IR Filter Shade 2.0 | N/A | N/A | N/A | |||
17-377-418
|
Honeywell™
S3300D |
Anti-fog, Scratch-resistant | Clear | N/A | N/A | N/A | |||
19-066-029
|
Honeywell™
S3300X |
Anti-fog | Clear | N/A | N/A | N/A | |||
19-066-031
|
Honeywell™
S3301 |
Scratch-resistant | Gray | N/A | N/A | N/A | |||
19-066-035
|
Honeywell™
S3302 |
Scratch-resistant | SCT-Reflect 60 | N/A | N/A | N/A | |||
19-066-036
|
Honeywell™
S3303X |
Anti-fog | SCT-Gray | N/A | N/A | N/A | |||
19-066-041
|
Honeywell™
S3308 |
Scratch-resistant | Mirror | N/A | N/A | N/A | |||
19-066-055
|
Honeywell™
S3300 |
Scratch-resistant | Clear | N/A | N/A | N/A | |||
19-185-707
|
Honeywell™
S3306HS |
Anti-fog | Shade 3 | N/A | N/A | N/A | |||
19-185-709
|
Honeywell™
S3310HS |
Anti-fog | Gray | N/A | N/A | N/A | |||
19-185-710
|
Honeywell™
S3311HS |
Anti-fog | SCT-Vermilion | N/A | N/A | N/A | |||
Description
- Soft, flexible nose fingers conform to fit a variety of nasal profiles
- Black frames with adjustable temples for comfortable fit; cushioned for all-day comfort
- Available lens tints include clear, gray, mirror, Infradura™ welding shades, and SCT™ to reduce spectral hazards
- All clear lenses absorb >99.9% of UVA and UVB radiation up to 385nm; tinted lenses absorb >99.9% UVA and UVB radiation up to 400nm
- Rx Insert available (see cat. no. 191300825)
uvextreme™ Anti-Fog Coating (XTR)
- Permanently bonded to lens
- Absorbs 99.9% of UV radiation
- Anti-scratch and -static
- Will not wear after repeated cleanings
- Permanently bonded to lens
- Scratch- and chemical-resistant
- ultra-dura coated exterior; uvextreme coated interior
- Lengths lens life
- Reduces glare for workers moving between indoor to outdoor work environments.
- Provide broad protection against accidental arcs. Equivalent to a Shade 2.0. welding filter, but not designed for any welding applications.
- Provide protection from short-term electric arcs and radiation generated when an arc accidentally occurs. Equivalent to a Shade 1.2 welding lens, but not designed for any welding applications.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Black |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title