Learn More
Honeywell Safety Products™ Uvex™ Hypershock™ Safety Eyewear
Catalog No. 19066217
Click to view available options
Frame Color:
Black
Brown
Clear
Clear Ice
Matte Black
Smoke Brown
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant, UV-treated
Hardcoat
Scratch-resistant
Lens Tint:
Amber
Blue Mirror
Clear
Espresso
Gold Mirror
Red Mirror
SCT-Blue
SCT-Reflect 50
This item is not returnable. View return policy
This item is not returnable. View return policy
Delivers ultimate combination of high-performance protection and aggressive sport inspired styling
    • Streamlined padded temples and molded nosepiece
    • Designed with anti-fog lens coating for long-lasting fog free vision
    • Provides 2X scratch-resistance
    • 100% dielectric
    • Integrated hinge mechanisms with dual cam action
    • Soft, molded inserts along temple provide comfortable and secure fit
    • Lightweight, wrap-around frame provides increased coverage, side protection and superior peripheral vision
Compliance
Meet ANSI Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Lens Tint SCT-Reflect 50
Product Type Safety Eyewear
For Use With (Application) Agriculture, Construction, Food Services, Manufacturing, Oil and Gas, Steel and Metals, Transportation, Wind Energy industries
Frame Color Smoke Brown
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.