Learn More
Honeywell Safety Products™ Uvex™ Hypershock™ Safety Eyewear
Catalog No. 19066212 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Brown
Clear
Clear Ice
Matte Black
Smoke Brown
Lens Tint:
Amber
Blue Mirror
Clear
Espresso
Gold Mirror
Red Mirror
SCT-Blue
SCT-Reflect 50
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant, UV-treated
Hardcoat
Scratch-resistant
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-066-212 Matte Black Blue Mirror -
19-066-222 Clear Ice Blue Mirror -
19-185-654 Matte Black Clear Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-066-221 Clear Ice Red Mirror Scratch-resistant
19-066-210 Matte Black SCT-Reflect 50 Scratch-resistant
19-066-213 Matte Black Espresso Hardcoat
19-066-217 Smoke Brown SCT-Reflect 50 Hardcoat
19-066-218 Smoke Brown Gold Mirror -
19-066-219 Smoke Brown Espresso -
19-185-719 Black Amber Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-185-720 Black SCT-Blue Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-185-722 Brown Espresso Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-185-723 Clear Clear Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-066-214 Matte Black SCT-Blue Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-066-211 Matte Black Red Mirror Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-066-212 Supplier Honeywell Safety Products™ Supplier No. S2945
Add to Cart
Add to Cart
This item is not returnable. View return policy
Delivers ultimate combination of high-performance protection and aggressive sport inspired styling
  • Streamlined padded temples and molded nosepiece
  • Designed with anti-fog lens coating for long-lasting fog free vision
  • Provides 2X scratch-resistance
  • 100% dielectric
  • Integrated hinge mechanisms with dual cam action
  • Soft, molded inserts along temple provide comfortable and secure fit
  • Lightweight, wrap-around frame provides increased coverage, side protection and superior peripheral vision
Applications:
Indoor and outdoor applications in construction, oil, gas and chemical, utilities, landscaping and manufacturing.
Compliance
Meet ANSI Z87.1-2010 standards.
Specifications
Lens Tint Blue Mirror
Product Type Safety Eyewear
For Use With (Application) Agriculture, Construction, Food Services, Manufacturing, Oil and Gas, Steel and Metals, Transportation, Wind Energy industries
Frame Color Matte Black