Learn More
Honeywell Safety Products™ Uvex™ Hypershock™ Safety Eyewear
Shop All Protective Industrial Products Inc. ProductsDescription
- Streamlined padded temples and molded nosepiece
- Designed with anti-fog lens coating for long-lasting fog free vision
- Provides 2X scratch-resistance
- 100% dielectric
- Integrated hinge mechanisms with dual cam action
- Soft, molded inserts along temple provide comfortable and secure fit
- Lightweight, wrap-around frame provides increased coverage, side protection and superior peripheral vision
Applications:
Indoor and outdoor applications in construction, oil, gas and chemical, utilities, landscaping and manufacturing.
Compliance
Meet ANSI Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Anti-fog, Anti-static, Scratch-resistant, UV-treated |
| Lens Tint | SCT-Blue |
| For Use With (Application) | Agriculture, Construction, Food Services, Manufacturing, Oil and Gas, Steel and Metals, Transportation, Wind Energy Industries |
| Frame Color | Black |