Learn More
Honeywell™ Uvex™ Livewire™ Sealed Eyewear
Catalog No. 19039923
Click to view available options
Lens Tint:
Amber
Clear
Espresso
Gray
SCT-Gray Lens
SCT-Low IR
SCT-Reflect 50
Frame Color:
Black
Matte Black
Silver
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant, UV-treated
Anti-fog, Scratch-resistant
Anti-static, Scratch-resistant, UV-treated
This item is not returnable. View return policy
Not for chemical splash applications
Catalog No. 19-039-923
Supplier: Honeywell™ S2602XP
This item is not returnable. View return policy
Not for chemical splash applications
For protecting workers against intense wind, sun, dirt, dust, debris, as well as high heat and humidity
  • Uvextreme Plus™ anti-fog lens coating with 10X longer-lasting anti-fog performance
  • Also available with Dura-streme dual anti-fog/anti-scratch coating
  • Foam-lined subframe
  • Gap-free seal around eyes to protect from debris
  • Flexible design can be worn with headband like goggle or with temples like spectacle
  • Padded temples provide ideal tension and flex for comfort fit
  • Ratcheting temples adjust lens angle by up to 15° for optimal coverage and air circulation
  • Adjustable, flame-resistant latex-free cloth headband
  • Wraparound frame design accommodates respirators, low-profile temples fits with earmuffs
  • Wide lens tint assortment available in sealed eyewear: Clear, Amber, Espresso, Gray, SCT-Reflect 50 (indoor/outdoor mirror), SCT-Gray, SCT-Low IR, Shade 3.0 and Shade 5.0
  • Available in Matte Black and Silver frame options
Certifications
CSA
Compliance
ANSI Z87.1-2010
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-static, Scratch-resistant, UV-treated
Lens Tint Amber
Product Type Sealed eyewear
For Use With (Application) Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy
Frame Color Matte Black
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.