Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex™ Mercury™ Safety Glasses
Reagent Grade,≥99%
Catalog Number | Mfr. No. | Lens Tint | Lens Coating | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Lens Coating | Price | Quantity & Availability | ||||
19-111-206
|
Honeywell™
S1500H5 |
Clear | N/A | N/A | N/A | ||||
19-111-207
|
Honeywell™
S1500X |
Clear | N/A | N/A | N/A | ||||
19-111-208
|
Honeywell™
S1501 |
Gray | N/A | N/A | N/A | ||||
19-111-209
|
Honeywell™
S1501X |
Gray | Anti-fog | N/A | N/A | N/A | |||
19-111-210
|
Honeywell™
S1502 |
Amber | N/A | N/A | N/A | ||||
19-111-212
|
Honeywell™
S1503 |
Light mirror | N/A | N/A | N/A | ||||
19-111-211
|
Honeywell™
S1502X |
Amber | N/A | N/A | N/A | ||||
19-111-214
|
Honeywell™
S1505 |
Blue Mirror | Hardcoat | N/A | N/A | N/A | |||
Description
- ≥95% for general laboratory use.
Specifications
Black, Gray |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title