Learn More
Honeywell™ Uvex™ Protege™ Safety Glasses
Catalog No. 191305864 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Sandstone
Lens Coating:
Impact-resistant
Impact-resistant, Scratch-resistant
Scratch-resistant
Lens Tint:
Clear
Gray
SCT-Reflect 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-130-5864 Sandstone Scratch-resistant Gray
19-130-5861 Black Impact-resistant, Scratch-resistant SCT-Reflect 50
19-130-5857 Black Impact-resistant, Scratch-resistant Clear
19-130-5860 Black Impact-resistant Gray
19-130-5865 Sandstone Impact-resistant Gray
19-130-5858 Black Impact-resistant Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-5864 Supplier Honeywell™ Supplier No. S4211
This item is not returnable. View return policy
Ultra-lightweight and soft temple tip pads for a secure, comfortable fit
  • Secure, wraparound brow accommodates a wide range of users
  • Proprietary Floating Lens™ design offers increased flexibility for better fit
  • Soft, clear nose pads prevent slipping
uvextra™ Anti-Fog Coating (AF)
  • Permanently bonded to lens
  • Minimizes fogging
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2015, Military V0 Ballistic Test
Lens Coating Scratch-resistant
Description Lifetime frame guarantee
Lens Tint Gray
Frame Color Sandstone
Temple Style Cushioned
Lens Material Polycarbonate