Learn More
Honeywell™ Uvex™ Protege™ Safety Glasses
Click to view available options
Frame Color:
Black
Sandstone
Lens Tint:
Clear
Gray
SCT-Reflect 50
Description
- Secure, wraparound brow accommodates a wide range of users
- Proprietary Floating Lens™ design offers increased flexibility for better fit
- Soft, clear nose pads prevent slipping
uvextra™ Anti-Fog Coating (AF)
- Permanently bonded to lens
- Minimizes fogging
- Permanently bonded to lens
- Scratch- and chemical-resistant
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Warranty and Services
Lifetime frame guarantee.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2015, CSA Z94.3-2015, Military V0 Ballistic Test |
| Lens Coating | Scratch-resistant |
| Description | Lifetime frame guarantee |
| Lens Tint | Clear |
| Frame Color | Black |
| Temple Style | Cushioned |
| Lens Material | Polycarbonate |