Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Catalog No. 19351456 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Metallic Blue
Lens Coating:
Anti-fog
Hardcoat
Scratch-resistant
Lens Tint:
Amber
Clear
Espresso
SCT-Reflect 50
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-351-456 Black Anti-fog SCT-Reflect 50
19-351-451 Black Scratch-resistant Clear
19-185-650 Black Anti-fog Clear
19-351-460 Metallic Blue Anti-fog SCT-Reflect 50
19-351-459 Metallic Blue - Amber
19-351-458 Metallic Blue - Clear
19-351-455 Black Anti-fog Amber
19-351-457 Metallic Blue Hardcoat Clear
19-351-454 Black Anti-fog Espresso
19-351-452 Black Anti-fog Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-351-456 Supplier Honeywell™ Supplier No. S0604X
This item is not returnable. View return policy
Eye protection against impact, sun, wind, dust and airborne debris
  • For dirty, high-particulate work environments
  • With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
  • Soft and flexible nasal pads
  • Soft, high-quality face cushioning and padded temple tips
  • Highly adjustable flame-resistant headband
  • Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meet ANSI Z87.1-2003.
Lens Tint SCT-Reflect 50
For Use With (Application) For applications like grinding, chipping, shaving and sanding
Frame Color Black
Temple Style Removable