Description
- For dirty, high-particulate work environments
- With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
- Soft and flexible nasal pads
- Soft, high-quality face cushioning and padded temple tips
- Highly adjustable flame-resistant headband
- Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Hardcoat |
| Certifications/Compliance | Meet ANSI Z87.1-2003. |
| Lens Tint | Clear |
| For Use With (Application) | For applications like grinding, chipping, shaving and sanding |
| Frame Color | Metallic Blue |
| Temple Style | Removable |