Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Catalog No. 19351459 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Metallic Blue
Lens Tint:
Amber
Clear
Espresso
SCT-Reflect 50
Lens Coating:
Anti-fog
Hardcoat
Scratch-resistant
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-351-459 Metallic Blue Amber -
19-351-451 Black Clear Scratch-resistant
19-185-650 Black Clear Anti-fog
19-351-460 Metallic Blue SCT-Reflect 50 Anti-fog
19-351-458 Metallic Blue Clear -
19-351-455 Black Amber Anti-fog
19-351-457 Metallic Blue Clear Hardcoat
19-351-456 Black SCT-Reflect 50 Anti-fog
19-351-454 Black Espresso Anti-fog
19-351-452 Black Clear Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-351-459 Supplier Honeywell™ Supplier No. S0622X
This item is not returnable. View return policy
Eye protection against impact, sun, wind, dust and airborne debris
  • For dirty, high-particulate work environments
  • With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
  • Soft and flexible nasal pads
  • Soft, high-quality face cushioning and padded temple tips
  • Highly adjustable flame-resistant headband
  • Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003.
Lens Tint Amber
For Use With (Application) For applications like grinding, chipping, shaving and sanding
Frame Color Metallic Blue
Temple Style Removable