Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear with Reading Magnifiers
Catalog No. 19351472 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-351-472 +2.00
19-351-473 +2.50
19-351-471 +1.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-351-472 Supplier Honeywell™ Supplier No. S0662X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Delivering unmatched protection against impact, wind, dust and debris, Uvex Seismic offers a range of diopter strengths for workers needing increased magnification for reading, inspection and other fine detail work.
The wraparound design provides exceptional coverage while precise placement of the magnifiers allow for distortion-free vision. Uvex Seismic Reading Magnifiers: powerful eyewear for today's demanding work environments.
  • Superior coverage ideal for dirty, high-particulate work environments such as grinding, chipping, shaving and sanding.
  • Available in Clear with Uvextra™ Anti-fog coating and three diopter strengths: +1.5. +2.0, and +2.5 magnifications.
  • Superior anti-fog coating, coupled with top and bottom indirect ventilation, minimizes fogging to keep workers safe and comfortable all day.
  • Flexible design can be worn with temples or with headband as conditions change.
  • Soft, flexible nasal pads conform to a variety of facial profiles and minimize slippage.
  • Soft, high-quality face cushioning and padded temple tips offer all-day comfort.
  • Highly adjustable flame-resistant headband provides excellent fit.
  • Replaceable flame-resistant, cushion-lined subframe and headband extend the life of the eyewear.
  • High impact protection certified to ANSI Z87+(High Impact) and tested by Honeywell to meet the requirements of the CSA Z94.3 standard. (ANSI-certified for use with and without cushion-lined frame).
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Sealed Eyewear with Reading Magnifiers
Frame Color Black
Magnification Power +2.00
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400