Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear
Catalog No. 19351460
Click to view available options
Frame Color:
Black
Metallic Blue
Lens Coating:
Anti-fog
Hardcoat
Scratch-resistant
Lens Tint:
Amber
Clear
Espresso
SCT-Reflect 50
This item is not returnable. View return policy
Catalog No. 19-351-460
Supplier: Honeywell™ S0624X
This item is not returnable. View return policy
Eye protection against impact, sun, wind, dust and airborne debris
  • For dirty, high-particulate work environments
  • With Dura-streme dual coating technology which combines two types of lens coatings - Uvextreme anti-fog (interior) and Ultra-dura anti-scratch (exterior), as well as Uvextra AF anti-fog/Hardcoat anti-scratch coating options
  • Soft and flexible nasal pads
  • Soft, high-quality face cushioning and padded temple tips
  • Highly adjustable flame-resistant headband
  • Replaceable flame-resistant, cushion-lined subframe
Applications:
For applications like grinding, chipping, shaving and sanding.
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint SCT-Reflect 50
For Use With (Application) For applications like grinding, chipping, shaving and sanding
Frame Color Metallic Blue
Temple Style Removable
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.