Learn More
Honeywell Safety Products™ Uvex™ Skyper™ Safety Glasses
Click to view available options
Lens Tint:
Clear
IR Filter Shade 2.0
IR Filter Shade 3.0
IR Filter Shade 5.0
Description
Custom fit with superior peripheral protection
- Polycarbonate lenses with single-lens design feature Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic coating
- Wraparound, impact resistant lenses block 99.9% of harmful UVA and UVB radiation
- Black frames with molded-in browguard reduce the danger of flying particulates
- Molded-in nosebridge offers comfort and safety in a variety of work applications
- Patented, adjustable, comfort-cushioned Duoflex™ temples
- Simple lens replacement system, ratcheting lens inclination
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-1999 standards.
Specifications
Specifications
| Certifications/Compliance | Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards. |
| Lens Coating | Scratch-resistant |
| Description | Shade 2.0 Infra-dura |
| Lens Tint | IR Filter Shade 2.0 |
| Frame Color | Black |
| Lens Material | Polycarbonate |