Description
- Ideal for impact, dust/airborne particles, chemical splash and optical radiation (UV and welding/heat radiation)
- Superior optics and panoramic field of vision enhanced with toric lens
- Pivoting clip and quick-adjusting wide headband
- Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
- Replacement lenses easily snap on
Order Info
Replacement parts and accessories available (see Cat. No.18-813-429).
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Type | dura-streme anti-fog/Scratch-resistant |
| Certifications/Compliance | ANSI Z87.1-2015, CSA Z94.3 |
| Lens Tint | Shade 5 |
| Product Type | OTG Goggles |
| For Use With (Application) | Cutting and gas welding operations |
| Frame Color | Navy |
| No. per Case | 50/Cs. |
| Frame Material | Neoprene |