Learn More
Honeywell Uvex™ Stealth Safety Goggles with Hydroshield™ Anti-Fog Lens Coating
Catalog No. 19018531 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Gray
Teal
Headband Material:
Neoprene
Lens Tint:
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Headband Material Lens Tint
19-018-531 Gray Neoprene Gray
19-018-530 Gray Neoprene Clear
19-185-794 Teal Neoprene Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-018-531 Supplier Honeywell Supplier No. S3961HS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Delivers consistent anti-fog protection that lasts 90x longer
  • Provides >180 seconds
  • Fog free time
  • Provides 2x scratch resistance
  • >99.9% UVA/UVB protection
  • Lifetime frame guarantee
  • Lens replacement lenses available
  • Indirect ventilation system
    • D3 certified
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Lens Coating Anti-fog
Lens Type HydroShield AF
Color Gray
Lens Tint Gray
Product Type Safety Goggles
Headband Material Neoprene