Learn More
Honeywell Uvex™ Stealth Safety Goggles with Hydroshield™ Anti-Fog Lens Coating
Delivers consistent anti-fog protection that lasts 90x longer
$55.80
Specifications
Lens Coating Anti-fog
Lens Type HydroShield AF
View More Specs
Products 3
Catalog Number Mfr. No. Color Headband Material Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Color Headband Material Lens Tint Price Quantity & Availability  
19-018-530
View Documents
Honeywell
S3960HS
Gray Neoprene Clear
Each for $55.80
Pack of 50 EA for $2,810.00
Only null left
Add to Cart
19-018-531
View Documents
Honeywell
S3961HS
Gray Neoprene Gray Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
19-185-794
View Documents
Honeywell
S39610HS
Teal Neoprene Clear Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
Description
Description
  • Provides >180 seconds
  • Fog free time
  • Provides 2x scratch resistance
  • >99.9% UVA/UVB protection
  • Lifetime frame guarantee
  • Lens replacement lenses available
  • Indirect ventilation system
    • D3 certified
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Specifications
Anti-fog
HydroShield AF
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.