Learn More
Honeywell™ Uvex™ Strategy™ Safety Goggles
Catalog No. 19167201 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Gray
Orange
Includes:
Fabric band
Neoprene band
Ventilation Details:
Closed vent
Direct vent
Indirect vent
Indirect vent with foam
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Includes Ventilation Details
19-167-201 Gray Fabric band Direct vent
19-167-203 Gray Neoprene band Indirect vent
19-167-202 Gray Neoprene band Closed vent
19-167-204 Gray Fabric band Indirect vent with foam
19-167-205 Orange Neoprene band Indirect vent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-167-201 Supplier Honeywell™ Supplier No. S3800
Add to Cart
Add to Cart
This item is not returnable. View return policy
Economical over-the-glass (OTG) goggle that delivers both premium comfort and maximum protection
Provide protection against impact, chemical splash, dust, sand and debris — even in extreme environments. Available in four models for a variety of work applications: direct vent, closed vent, indirect vent and indirect vent with foam lining for fine particle protection.
  • Ideal for most indoor work applications
  • Rugged design that withstands the harshest environments
  • Lens tint: clear
  • Lenses offer 99.9% ultra-violet protection
  • Headband available in fabric or neoprene for chemical applications
  • Uvextra™ anti-fog coating minimizes fogging
  • Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection
  • Fits comfortably over most prescription eyewear
  • Economically priced
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Lens Type Uvextra
Lens Tint Clear
Product Type Safety goggles
Frame Color Gray
Includes Fabric band
Ventilation Details Direct vent