Description
Provide protection against impact, chemical splash, dust, sand and debris — even in extreme environments. Available in four models for a variety of work applications: direct vent, closed vent, indirect vent and indirect vent with foam lining for fine particle protection.
- Ideal for most indoor work applications
- Rugged design that withstands the harshest environments
- Lens tint: clear
- Lenses offer 99.9% ultra-violet protection
- Headband available in fabric or neoprene for chemical applications
- Uvextra™ anti-fog coating minimizes fogging
- Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection
- Fits comfortably over most prescription eyewear
- Economically priced
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Type | Uvextra |
| Lens Tint | Clear |
| Product Type | Safety goggles |
| Frame Color | Gray |
| Includes | Neoprene band |
| Ventilation Details | Closed vent |