Learn More
Honeywell™ Uvex™ Strategy™ Safety Goggles
Catalog No. 19167203
Click to view available options
Frame Color:
Gray
Orange
Includes:
Fabric band
Neoprene band
Ventilation Details:
Closed vent
Direct vent
Indirect vent
Indirect vent with foam
Economical over-the-glass (OTG) goggle that delivers both premium comfort and maximum protection
Provide protection against impact, chemical splash, dust, sand and debris — even in extreme environments. Available in four models for a variety of work applications: direct vent, closed vent, indirect vent and indirect vent with foam lining for fine particle protection.
  • Ideal for most indoor work applications
  • Rugged design that withstands the harshest environments
  • Lens tint: clear
  • Lenses offer 99.9% ultra-violet protection
  • Headband available in fabric or neoprene for chemical applications
  • Uvextra™ anti-fog coating minimizes fogging
  • Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection
  • Fits comfortably over most prescription eyewear
  • Economically priced
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type Uvextra
Lens Tint Clear
Product Type Safety Goggles
Frame Color Gray
Includes Neoprene band
Ventilation Details Indirect vent
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.