Learn More
Honeywell™ Uvex™ SVP 200 Series
Catalog No. 19039964 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Blue
Clear
Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color
19-039-964 Blue
19-039-963 Clear
19-039-965 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-039-964 Supplier Honeywell™ Supplier No. SVP205
Add to Cart
Add to Cart
This item is not returnable. View return policy
Honeywell™ Uvex™ SVP 200 Series offers stylish, low-profile and very lightweight fit for all day comfort.
  • PC lens, HC, Uvextra AF
  • Stylish, VERY lightweight, 10-base wrap-around frame
  • Flexible tips
  • Molded Nosepiece
  • Available in many lens tints
Specifications
Lens Coating Anti-fog
Width (Metric) 6.6 cm
Certifications/Compliance ANSI Z.87.1 2015, CSA Z94.3 standard
Lens Tint Clear
For Use With (Application) Eye protection
Frame Color Blue
Model UVEX SVP205
No. per Pack 10/Pk.
Temple Style Spatula
Frame Material Polycarbonate
UV Protection U6