Learn More
Honeywell™ Uvex™ Ultraspec™ 2001 OTG Safety Glasses
Catalog No. 19034243 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Clear
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-034-243 Scratch-resistant Gray
19-034-241 Scratch-resistant Clear
19-034-242 Anti-fog Clear
19-034-244 Anti-fog Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-034-243 Supplier Honeywell™ Supplier No. S0113
This item is not returnable. View return policy
Comfortable fit over prescription eyewear
These lightweight, unilens spectacles with built-in browguard fit easily over prescription eyewear. Adjustable length Duoflex™ temples provide less pressure behind the ear for improved comfort. In addition, both clear and gray tinted lenses are available with Uvextreme™ coating that resists scratching, fogging, and static. Gray lens is appropriate for outdoor wear; both clear and gray lenses absorb 99.9% of UV radiation up to 400nm.
Order Info
Spectacles weigh 51g.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Type Ultra-dura Hardcoat
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2010
Lens Tint Gray
Product Type Eyewear
Temple Style Duoflex
Weight (Metric) 51 g
UV Protection 0.999