Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Clear UV Blocking Covers

Catalog No. UVP19011201 Shop All UVP Inc. Products
Change view
Click to view available options
For Use With (Equipment):
3uv™ transilluminators
Benchtop transilluminators
Standard Format UV Transilluminators
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
UVP19011201 Standard Format UV Transilluminators
UVP19012101 3uv™ transilluminators
UVP19012001 Benchtop transilluminators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. UVP19011201 Supplier UVP Supplier No. 19011201

Specifications

For Use With (Application) Gel electrophoresis
Product Type UV Blocking Cover
Quantity each
Color Clear
For Use With (Equipment) Standard Format UV Transilluminators
Includes Uv Blocking Cover
Size 18 x 13 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.