Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Gel Tools: Gel Ruler

Catalog No. UVP85000301 Shop All UVP Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Transilluminator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
UVP85000301 Transilluminator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP85000301 Supplier UVP Supplier No. 85000301
Only null left
Add to Cart
Add to Cart

Provide reference marks for DNA analysis

  • Contain centimeter markings that fluoresce under 365nm and 302nm ultraviolet wavelengths

Specifications

For Use With (Equipment) Transilluminator
Type Electrophoresis Fluorescent Gel Ruler
Includes Centimeter marking
Length (English) 9.84 in.
Length (Metric) 25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.