Learn More
UVP J-Series Photovoltaic UV Intensity Meters: Filter Contrast
Catalog No. UVP38004301 Shop All UVP Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Models J-221 and J-225 analog UV Intensity Meters
Catalog No. For Use With (Equipment)
UVP38004301 Models J-221 and J-225 analog UV Intensity Meters
1 options
Catalog No. UVP38004301 Supplier UVP Supplier No. 38004301
Replacement part for Photovoltaic J-Series UV meters
All meters and sensors are individually serialized for traceability of calibration. Calibration is recommended every six months and is certified to meet UVP's published standards and is traceable to NIST. Calibrations are performed at UVP's factory.
Quality control
Specifications
Type Filter Contrast
For Use With (Equipment) Models J-221 and J-225 analog UV Intensity Meters