Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP PhotoDoc-It 60 Imaging Systems
SCI_ED

Catalog No. S99544 Shop All UVP Inc. Products
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
S99544 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99544 Supplier UVP Supplier No. 97027401/EMD

Capture, print and save color images for basic gel documentation with this compact, economical laboratory package.

  • DigiCam 60 color camera with 10.0 megapixel resolution
  • Lightweight, compact hood provides a compact darkroom environment, hood now includes a new UV blocking viewer window
  • Benchtop UV Transilluminator delivers back-lit UV; select from several benchtop models
  • Color printer generates instant 4 x 6 in. low cost, laboratory quality color prints
  • Ethidium bromide filter for fluorescent stained gels
  • Combination lock secures the camera to the darkroom

Specifications

Voltage 115 V
Transilluminator No transilluminator
Camera Type DigiCam 60 w/10.0 megapixel resolution
Product Type PhotoDoc-It 60 Imaging System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.