Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Power Supply for Portable UV Lamps

Catalog No. UVP75000201 Shop All UVP Inc. Products
Change view
Click to view available options
Wattage:
6 W
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wattage
UVP75000201 6 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP75000201 Supplier UVP Supplier No. 75000201
Only null left
Add to Cart
Add to Cart

Specifications

Voltage 12 VDC
Wattage 6 W
For Use With (Equipment) UVG47, ML49, UVGL48 portable UV lamp
Includes Power Supply Board
Packaging Quantity 1 Ea.
Product Type Power Supply Board
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.