Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UVRAG Antibody (953035), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9180N
Description
UVRAG Monoclonal antibody specifically detects UVRAG in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
UVRAG | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
953035 | |
Flow Cytometry, CyTOF-ready | |
beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38 | |
Human UVRAG synthetic peptide corresponding to sequences between amino acids 260-360, Accession # Q9P2Y5 | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Cellular Signaling, Epigenetics | |
7405 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction