Learn More
Invitrogen™ UVRAG Polyclonal Antibody

Description
UVRAG Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | UVRAG |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Uvrag |
| Gene Accession No. | Q8K245, Q9P2Y5 |
| Gene Alias | 9530039D02Rik; AI648770; BB124205; beclin 1 binding protein; DHTX; disrupted in heterotaxy; hypothetical protein; p63; UV radiation resistance associated; UV radiation resistance associated gene; UV radiation resistance-associated gene protein; UV radiation resistance-associated protein; UVRAG; Uvrag1; Uvragl; VPS38 |
| Gene Symbols | Uvrag |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.