Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UVRAG Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
Supplier: Leinco Technologies Inc 23013100UG

This item is not returnable.
View return policy
Description
UVRAG Polyclonal antibody specifically detects UVRAG in Human, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
UVRAG | |
Polyclonal | |
Unconjugated | |
Uvrag | |
9530039D02Rik; AI648770; BB124205; beclin 1 binding protein; DHTX; disrupted in heterotaxy; hypothetical protein; p63; UV radiation resistance associated; UV radiation resistance associated gene; UV radiation resistance-associated gene protein; UV radiation resistance-associated protein; UVRAG; Uvrag1; Uvragl; VPS38 | |
Synthetic peptide corresponding to mid-protein amino acids of human UVRAG. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q9P2Y5 | |
Rabbit | |
Protein A | |
RUO | |
308846, 7405 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction