Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UVSX Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144482 Supplier Invitrogen™ Supplier No. PA5144482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UVSX Polyclonal Antibody for Western Blot, ELISA

Important in genetic recombination, DNA repair, and replication. Possesses pairing and strand-transfer activity. Interacts with dda and gene 32 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UVSX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene UVSX
Gene Accession No. P04529
Gene Alias Recombination and repair protein UVSX
Gene Symbols UVSX
Host Species Rabbit
Immunogen Recombinant Enterobacteria phage T4 (Bacteriophage T4) UVSX protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1258704
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.