Learn More
Invitrogen™ UXT Polyclonal Antibody

Description
Immunogen sequence: LKFIDRKSSL LTELSNSLTK DSMNIKAHIH MLLEGLRELQ GLQNFPEKPH H Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
Specifications
Specifications
| Antigen | UXT |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UXT |
| Gene Accession No. | Q9UBK9 |
| Gene Alias | 0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT |
| Gene Symbols | UXT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.